Phosphate-buffered saline (PBS, 10X), pH 7,4
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00131855
- MDL No.:MFCD00131855
- Physical form:Liquid
- Appearance:Colourless
Frequently Bought Together
Physical Form: Liquid
Appearance: Colourless
MDL No.: MFCD00131855