Order Entry
Austria
ContactUsLinkComponent
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M in aqueous solution, autoclaved pH 8.0
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M in aqueous solution, autoclaved pH 8.0
Catalog # J61721.AD
CAS Number:  
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M in aqueous solution, autoclaved pH 8.0
Catalog # J61721.AD
CAS Number:  

Specifications

  • Pk:
    50 mL

Specifications

About this item

 :
 :380.35 g/mol
Storage Temperature:Ambient
CAS Number:67-42-5
Formula:C₁₄H₂₄N₂O₁₀
MDL Number:MFCD00004291