Adenosine 5'-monophosphate disodium salt (AMP disodium salt) hydrate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4578-31-8
:
Formula:C₁₀H₁₂N₅Na₂O₇P·H₂O
MDL Number:MFCD00065023
Merck Index:14,158
:391.2 g/mol (anhy)
Storage Temperature:Freezer
EINECS:224-961-2
Related Information
Frequently Bought Together