CHES, 0.5 M buffer solutions
Supplier: Thermo Fisher Scientific
Certificates
About this item
2-(Cyclohexylamino)ethanesulfonic acid, CHES.
CAS No.: 103-47-9
MDL No.: MFCD00003835
Specifications
- CAS No.:103-47-9
- MDL No.:MFCD00003835
- Storage temperature:2°C to 8°C
- Physical form:Liquid
Specifications
Frequently Bought Together