
Polyethyleneimine, M.W. 60,000 50% (w/w) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9002-98-6
:
Formula:(CH2CH2NH)n
MDL Number:MFCD00084427
Density:1.07
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together