Order Entry
Austria
ContactUsLinkComponent
Stripping buffer (4X)
Stripping buffer (4X)
Catalog # J60925.AP
CAS Number:  
Stripping buffer (4X)
Catalog # J60925.AP
CAS Number:  

Specifications

  • Pk:
    500 mL
  • Description:
    Stripping buffer, 4X
  • Physical form:
    Colourless and odourless liquid

Specifications

About this item

Physical Form: Liquid
Appearance: Colourless
Odour: Odourless