TRIS-buffered saline (TBS, 20X) pH 7,4
CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00132476
- CAS No.:77-86-1
- MDL No.:MFCD00132476
- Physical form:Liquid
Frequently Bought Together
CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00132476