Order Entry
Austria
ContactUsLinkComponent
Phosphate-buffered saline (PBS, 10X), pH 7,4, for Western blot
Phosphate-buffered saline (PBS, 10X), pH 7,4, for Western blot
Certificates
Phosphate-buffered saline (PBS, 10X), pH 7,4, for Western blot
Certificates

Specifications

  • MDL No.:
    MFCD00131855
  • Physical form:
    Liquid
  • Appearance:
    Colourless

Specifications

Related Information

Frequently Bought Together