TRIS-buffered saline (TBS, 20X), with 2% Tween 20
Supplier: Thermo Fisher Scientific
Certificates
About this item
Physical Form: Liquid
MDL No.: MFCD00678198
Specifications
- MDL No.:MFCD00678198
- Physical form:Liquid
Specifications
Related Information
Frequently Bought Together