Buffers, BICINE, 0,5 M solutioins
Supplier: Thermo Fisher Scientific
Certificates
About this item
N,N-(2-Dihydroxyethyl)glycine (BICINE) buffer solutions.
CAS No.: 150-25-4
MDL No.: MFCD00004295
BICINE is used for the preparation of stable substrate solution for serum guanase determination. The use of bicine in a thin layer ion exchange chromatography method for protein resolution has been published. Bicine has been utilized in peptide and protein crystallization. A kinetic study of a quaternary transition-state analogue complex of creatine kinase used bicine in the reaction buffer. A multiphasic buffer system for SDS-PAGE of proteins and peptides that incorporates bicine has been described.
Caution: Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
- CAS No.:150-25-4
- MDL No.:MFCD00004295
- Storage temperature:2°C to 8°C
- Physical form:Liquid
Specifications
Related Information
Frequently Bought Together