TRIS-buffered saline (TBS, 10X), with 0,5% Tween 20
Physical Form: Viscous liquid
Appearance: Colourless
MDL No.: MFCD00678198
- MDL No.:MFCD00678198
- Physical form:Viscous liquid
- Appearance:Colourless
Frequently Bought Together
Physical Form: Viscous liquid
Appearance: Colourless
MDL No.: MFCD00678198