Formaldehyde 4% in PBS stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
ADR:8,III
:Stabilised with methanol
:GHS08
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together