TAPS, 0,2 M buffer soln., pH 8,0
CAS No.: 91000-53-2
Physical Form: Liquid
MDL No.: MFCD00069903
- CAS No.:91000-53-2
- MDL No.:MFCD00069903
- Physical form:Liquid
Frequently Bought Together
CAS No.: 91000-53-2
Physical Form: Liquid
MDL No.: MFCD00069903