Glycine, 0.2M buffer soln., pH 3.5
CAS No.: 56-40-6
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00008131
- CAS No.:56-40-6
- MDL No.:MFCD00008131
- Physical form:Liquid
- Odour/scent:Odourless
Frequently Bought Together
CAS No.: 56-40-6
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00008131