Order Entry
Austria
ContactUsLinkComponent
Transformation reagent, TB solution-II
Transformation reagent, TB solution-II
Catalog # J60028.AK
CAS Number:  
Transformation reagent, TB solution-II
Catalog # J60028.AK
CAS Number:  

Specifications

  • Pk:
    250 mL
  • Description:
    TB Solution-II
  • Storage conditions:
    2°C to 8°C

Specifications

About this item

For transformation of competent cells.

Contains 250 mM potassium chloride, 15 mM calcium chloride, 10 mM HEPES, 55 mM manganese chloride, pH 6,5.

Applications: For transformation of competent cells.