Vitamin A acetate in gelatin, 500,000 I.U./g
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:127-47-9
:
Formula:C₂₂H₃₂O₂
MDL Number:MFCD00019413
:328.49 g/mol
Merck Index:14,10013
EINECS:204-844-2
:GHS08
:GHS07
Storage Temperature:Refrigerator
Related Information
Frequently Bought Together