
3,6-Dihydro-2H-pyridine-1-carboxylic acid tert-butyl ester 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:85838-94-4
:
Formula:C₁₀H₁₇NO₂
MDL Number:MFCD04972245
:183.25 g/mol
Boiling Pt:78…84 °C (5 mmHg)
Density:1.029
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together