
(+)-Bis[(R)-1-phenylethyl]amine ≥99%, ee 98%, ChiPros®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23294-41-9
:
Formula:C₁₆H₁₉N
MDL Number:MFCD00243088
:225.34 g/mol
Boiling Pt:98 °C (0.3 mmHg)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together