Chlorpheniramine maleate 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:113-92-8
:
Formula:C₁₆H₁₉ClN₂
MDL Number:MFCD00069225
:390.86 g/mol
Melting Pt:130…135 °C
EINECS:204-037-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together