Rose Bengal
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:632-69-9
:MFCD00005043
Formula:C₂₀H₂Cl₄I₄O₅Na₂
MDL Number:MFCD00151169
:1017.63 g/mol
Merck Index:14,8262
Melting Pt:>300 °C
EINECS:211-183-3
:GHS07
Storage Temperature:Ambient
Frequently Bought Together