
Trisodium 2-(4-Sulphophenylazo)-1,8-dihydroxynaphthalene-3,6-disulphonate (SPADNS)
Catalog # A16933.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:570.40 g/mol
Melting Pt:>300 °C
EINECS:245-803-9
:GHS07
Storage Temperature:Ambient
CAS Number:23647-14-5
Formula:C₁₆H₁₂N₂O₁₁S₃
MDL Number:MFCD00003952