Polyethylene, powder 500 micron, low density
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:9002-88-4
:
Formula:(CH₂CH₂)n
MDL Number:MFCD00084423
Merck Index:14,7567
Density:0.92
Storage Temperature:Ambient
Related Information
Frequently Bought Together