Order Entry
Austria
ContactUsLinkComponent
Buffer Solutions
Buffer Solutions
Certificates
Buffer Solutions
Certificates

Specifications

  • MDL No.:
    MFCD00147253
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Appearance:
    Colourless

Specifications

Frequently Bought Together