Thallium 203, 203 Tl, plasma standard solution, 203 Tl 10 µg/ml, Specpure®
Catalog # 45288.AD
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:2% HNO₃
- Pk:50 mL
- MDL No.:MFCD00084592
- Element:Thallium
- Symbol:Tl
Specifications
About this item
Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00084592