Order Entry
Austria
ContactUsLinkComponent
Selenium 82, 82 Se, plasma standard solution, 82 Se 10 µg/ml, Specpure®
Selenium 82, 82 Se, plasma standard solution, 82 Se 10 µg/ml, Specpure®
Catalog # 45284.AD
CAS Number:  
Selenium 82, 82 Se, plasma standard solution, 82 Se 10 µg/ml, Specpure®
Catalog # 45284.AD
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    50 mL
  • MDL No.:
    MFCD00084581
  • Element:
    Selenium
  • Symbol:
    Se

Specifications

About this item

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00084581