Order Entry
Austria
ContactUsLinkComponent
Cadmium 106, 106 Cd, plasma standard solution, 106 Cd 10 µg/ml, Specpure®
Cadmium 106, 106 Cd, plasma standard solution, 106 Cd 10 µg/ml, Specpure®
Catalog # 45274.AD
CAS Number:  
Cadmium 106, 106 Cd, plasma standard solution, 106 Cd 10 µg/ml, Specpure®
Catalog # 45274.AD
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    50 mL
  • MDL No.:
    MFCD00143885
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item

Molecular Formula: Matrix: 2% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD00143885