Order Entry
Austria
ContactUsLinkComponent
Polyethylene, sheets, low density, Thickness: 3.18 mm (0.125 in)
Polyethylene, sheets, low density, Thickness: 3.18 mm (0.125 in)
Catalog # 45175.HB
CAS Number:  
Polyethylene, sheets, low density, Thickness: 3.18 mm (0.125 in)
Catalog # 45175.HB
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
Density:0.92
Storage Temperature:Ambient
CAS Number:9002-88-4
Formula:(CH₂CH₂)n
MDL Number:MFCD00084423