Order Entry
Austria
ContactUsLinkComponent
Instrument calibration(ICAL) standard, Solution A, Specpure®
Instrument calibration(ICAL) standard, Solution A, Specpure®
Catalog # 44855.AE
CAS Number:  
Instrument calibration(ICAL) standard, Solution A, Specpure®
Catalog # 44855.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Instrument Calibration(ICAL) Standard, Solution A
  • MDL No.:
    MFCD08064526

Specifications

About this item

Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD08064526

Concentration:
Ag 200 µg/ml ±2%
Ba 1,000 µg/ml ±2%
Be 400 µg/ml ±2%
Cd 500 µg/ml ±2%
Co 1,000 µg/ml ±2%
Cu 1,000 µg/ml ±2%
Fe 1,000 µg/ml ±2%
Mn 1,000 µg/ml ±2%
Ni 1,000 µg/ml ±2%
Pb 1,000 µg/ml ±2%
Tl 1,000 µg/ml ±2%
Zn 1,000 µg/ml ±2%