Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Pk:100 mL
- Description:ICP-MS Internal Standard solution for 200.8 & CLP ILM05.2, Specpure®
- MDL No.:MFCD03792350
Specifications
About this item
These standards are applicable for the determination of elements as dissolved elements in ground waters, surface waters, and drinking water, and total recoverable elements in these waters as well as wastewaters, sludges, and soils samples.
Matrix: 5% HNO₃
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD03792350
Concentration: Bi 10 µg/ml ±2%; In 10 µg/ml ±2%; Lu 10 µg/ml ±2%; Sc 10 µg/ml; ±2%; Tb 10 µg/ml ±2%; Y 10 µg/ml ±2%; ₆Li 10 µg/ml ±2%