Order Entry
Austria
ContactUsLinkComponent
ICP-MS internal standard solution, Specpure®
ICP-MS internal standard solution, Specpure®
Catalog # 44651.AE
CAS Number:  
ICP-MS internal standard solution, Specpure®
Catalog # 44651.AE
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    8 component standard, 100 µg/ml
  • MDL No.:
    MFCD00803321

Specifications

About this item

UN No.: UN3264
MDL No.: MFCD00803321

Concentration:
Bi 100 µg/ml ±2%;
Ga 100 µg/ml ±2%;
In 100 µg/ml ±2%;
Sc 100 µg/ml ±2%;
Tb 100 µg/ml ±2%;
Y 100 µg/ml ±2%;
6Li 100 µg/ml ±2%