Order Entry
Austria
Orders LinkContactUsLinkComponent
Buffer HF-Improved

Buffer HF-Improved

Buffer HF-Improved
Certificates
Buffer HF-Improved

2 Options Available Below

Product Details & Documents

Buffer used for pretreatment of planar silicon devices when plating with nickel plating solution products 44069 and 44070.

Density: 1.12
Appearance: Colorless
Odor: Acrid
Boiling Point: 103…105°
UN No.: 2817

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 44627.AK
  • 44627.K2
  • Odour/scent:
  • Acrid
  • Acrid
  • MDL No.:
  • MFCD00132734
  • MFCD00132734
  • Appearance:
  • Colourless
  • Colourless
  • Density:
  • 1.12 g/mL
  • 1.12 g/mL
  • UN No.:
  • 2817
  • 2817

Specifications

Product Family Options

Product Information

  • Pk
    Availability
    Price
  • Buffer HF-Improved
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 41,40 €
    41,40 €
    1 * 250 mL
       41,40 €

    Specifications:
    More
    Specifications
    Cat. No.
    44627.AK
    Odour/scent
    Acrid
    MDL No.
    MFCD00132734
    Appearance
    Colourless
    Density
    1.12 g/mL
    UN No.
    2817
  • Buffer HF-Improved
    1 L
    Log in for availability
    Log in for availability
    1 * 1 L
    1 * 1 L - 118,00 €
    118,00 €
    1 * 1 L
       118,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    44627.K2
    Odour/scent
    Acrid
    MDL No.
    MFCD00132734
    Appearance
    Colourless
    Density
    1.12 g/mL
    UN No.
    2817