Order Entry
Austria
ContactUsLinkComponent
 
ICP-MS Water spike standard solution for 6020 CLP-M, Specpure®
Catalog # 44539.AE
CAS Number:  
ICP-MS Water spike standard solution for 6020 CLP-M, Specpure®
Catalog # 44539.AE
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃/tr. (CHOH)₂(COOH)₂
  • Pk:
    100 mL
  • Description:
    ICP-MS Water Spike Standard Solution for 6020 CLP-M
  • MDL No.:
    MFCD02266368

Specifications

About this item

Molecular Formula: Matrix: 5% HNO₃/tr. tartaric acid
Physical Form: Liquid
UN No.: UN3264
MDL No.: MFCD02266368

Concentration:
Ag 10 µg/ml ±2%
Al 300 µg/ml ±2%
As 10 µg/ml ±2%
Ba 100 µg/ml ±2%
Be 10 µg/ml ±2%
Cd 10 µg/ml ±2%
Co 5 µg/ml ±5%
Cr 20 µg/ml ±2%
Cu 20 µg/ml ±2%
Mn 5 µg/ml ±5%
Ni 10 µg/ml ±2%
Pb 10 µg/ml ±2%
Sb 20 µg/ml ±2%
Se 50 µg/ml ±2%
Tl 10 µg/ml ±2%
V 10 µg/ml ±2%
Zn 10 µg/ml ±2%