Transition metals, plasma standard solution, Specpure®
Supplier: Thermo Fisher Scientific
Transition metals, plasma standard solution, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Cd, Co, Cr, Cu, Fe, Mn, Ni, V, Zn @ 100 µg/ml
UN No.: UN3264
MDL No.: MFCD00151266
2 Options Available Below
Product Details & Documents
Cd, Co, Cr, Cu, Fe, Mn, Ni, V, Zn @ 100 µg/ml
UN No.: UN3264
MDL No.: MFCD00151266
Documents
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 44518.AE
- 44518.AP
- MDL No.:
- MFCD00151266
- MFCD00151266
Specifications
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



