Order Entry
Austria
ContactUsLinkComponent
Multi-element plasma standard solution 4, Specpure®
Multi-element plasma standard solution 4, Specpure®
Catalog # 42885.AE
CAS Number:  
Multi-element plasma standard solution 4, Specpure®
Catalog # 42885.AE
CAS Number:  

Specifications

  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Description:
    Multi-Element Plasma Standard Solution 4
  • MDL No.:
    MFCD00212340

Specifications

About this item

UN No.: UN3264
MDL No.: MFCD00212340