Order Entry
Austria
ContactUsLinkComponent
Multi-element plasma standard solution 2, Specpure®
Multi-element plasma standard solution 2, Specpure®
  42882.AE
 :  
Multi-element plasma standard solution 2, Specpure®
  42882.AE
 :  

 

  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Description:
    Multi-Element Plasma Standard Solution 2
  • MDL No.:
    MFCD00212338

 

 

UN No.: UN3264
MDL No.: MFCD00212338