Order Entry
Austria
ContactUsLinkComponent
 
Multi-element plasma standard solution 1, Specpure®
  42881.AE
 :  
Multi-element plasma standard solution 1, Specpure®
  42881.AE
 :  

 

  • Matrix:
    Nitric acid dil.
  • Pk:
    100 mL
  • Description:
    Multi-Element Plasma Standard Solution 1
  • MDL No.:
    MFCD00212337

 

 

UN No.: UN3264
MDL No.: MFCD00212337