Order Entry
Austria
ContactUsLinkComponent
ICP-MS analytes B for 6020 CLP-M, Specpure®
ICP-MS analytes B for 6020 CLP-M, Specpure®
Catalog # 42650.AE
CAS Number:  
ICP-MS analytes B for 6020 CLP-M, Specpure®
Catalog # 42650.AE
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Description:
    ICP-MS Analytes B for 6020 CLP-M
  • MDL No.:
    MFCD02262473

Specifications

About this item

11 elements

UN No.: UN3264
MDL No.: MFCD02262473

Concentration:
Ag 5 µg/ml ±5%
As 10 µg/ml ±2%
Cd 10 µg/ml ±2%
Co 20 µg/ml ±2%
Cr 20 µg/ml ±2%
Cu 20 µg/ml ±2%
Mn 20 µg/ml ±2%
Ni 20 µg/ml ±2%
Se 10 µg/ml ±2%
V 20 µg/ml ±2%
Zn 10 µg/ml ±2%