Order Entry
Austria
ContactUsLinkComponent
Oxalate, Ion chromatography standard solution, Specpure®, C_2O4-2 1000 µg/ml
Oxalate, Ion chromatography standard solution, Specpure®, C_2O4-2 1000 µg/ml
Catalog # 42568.AE
CAS Number:  
Oxalate, Ion chromatography standard solution, Specpure®, C_2O4-2 1000 µg/ml
Catalog # 42568.AE
CAS Number:  

Specifications

  • Concentration:
    1000 mg/l
  • Pk:
    100 mL
  • Description:
    Oxalate standard solution
  • MDL No.:
    MFCD00012465
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Molecular formula:
    Na2C2O4 in H2O
  • Contents:
    Na₂C₂O₄ in H₂O

Specifications

About this item

Molecular Formula: Na₂C₂O₄ in H₂O
Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00012465