Specpure® Silica, Standard Solution, SiO₂ 1000 µg/ml
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Na₂SiO₃ in H₂O
Physical Form: Liquid
Specifications
- MDL No.:MFCD00011232
- Physical form:Liquid
- Molecular formula:Na₂SiO₃ in H₂O
Specifications
Related Information
Frequently Bought Together