Order Entry
Austria
ContactUsLinkComponent
Standards, sulphur in light mineral oil standard solution, Specpure®
Standards, sulphur in light mineral oil standard solution, Specpure®
Certificates
Standards, sulphur in light mineral oil standard solution, Specpure®
Certificates

Specifications

  • MDL No.:
    MFCD00804270
  • Physical form:
    Liquid

Specifications

Frequently Bought Together