Eicosanedioic acid
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2424-92-2
:
Formula:C₂₀H₃₈O₄
MDL Number:MFCD00059627
Melting Pt:127 °C
:342.52 g/mol
Storage Temperature:Ambient
Related Information
Frequently Bought Together