Order Entry
Austria
ContactUsLinkComponent
Sulfur in residual oil standard solutions, Specpure®
Sulfur in residual oil standard solutions, Specpure®
Certificates
Sulfur in residual oil standard solutions, Specpure®
Certificates

Specifications

  • MDL No.:
    MFCD00197838
  • Physical form:
    Liquid

Specifications

Frequently Bought Together