Sulphur in kerosene standard solutions, Specpure®
Specpure® calibration standards, designed specifically for the analysis of sulphur in petroleum products by X-ray fluorescence spectrometry.
Physical Form: Liquid
UN No.: UN1223
MDL No.: MFCD00804268
- MDL No.:MFCD00804268
- UN No.:1223
- Physical form:Liquid
Frequently Bought Together