Order Entry
Austria
ContactUsLinkComponent
ICP-MS stock standard solution B for 200,8, Specpure®
ICP-MS stock standard solution B for 200,8, Specpure®
Catalog # 40486.AE
CAS Number:  
ICP-MS stock standard solution B for 200,8, Specpure®
Catalog # 40486.AE
CAS Number:  

Specifications

  • Matrix:
    1% HNO₃
  • Pk:
    100 mL
  • Description:
    ICP-MS Stock Standard solution B
  • MDL No.:
    MFCD00803327

Specifications

About this item