Order Entry
Austria
ContactUsLinkComponent
ICP-MS tuning standard solution for 6020 CLP-M, Specpure®
  40482.AE
 :  
ICP-MS tuning standard solution for 6020 CLP-M, Specpure®
  40482.AE
 :  

 

  • Matrix:
    2% HNO₃
  • Pk:
    100 mL
  • Description:
    ICP-MS Tuning Standard Solution for 6020 CLP-M
  • MDL No.:
    MFCD00240453

 

 

UN No.: UN3264
MDL No.: MFCD00240453