Order Entry
Austria
ContactUsLinkComponent
Multi Ion anion IC standard solution, Specpure® (Dionex) in H₂O
Multi Ion anion IC standard solution, Specpure® (Dionex) in H₂O
Catalog # 35565.AE
CAS Number:  
Multi Ion anion IC standard solution, Specpure® (Dionex) in H₂O
Catalog # 35565.AE
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    Multi Ion anion IC standard solution, Specpure® (Dionex) in H₂O
  • MDL No.:
    MFCD00144813
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless

Specifications

About this item

Physical Form: Liquid
Odour: Odourless
MDL No.: MFCD00144813