Order Entry
Austria
ContactUsLinkComponent
 
Bromide, Ion chromatography standard solution, Specpure®, Br- 1000µg/ml
 
Bromide, Ion chromatography standard solution, Specpure®, Br- 1000µg/ml
 

 

  • MDL No.:
    MFCD00134172
  • Physical form:
    Liquid
  • Molecular formula:
    KBr in H2O

 

Frequently Bought Together