Order Entry
Austria
ContactUsLinkComponent
 
Chloride, Ion chromatography standard solution, Specpure®, Cl- 1000µg/ml
 
Chloride, Ion chromatography standard solution, Specpure®, Cl- 1000µg/ml
 

 

  • MDL No.:
    MFCD00143949
  • Physical form:
    Liquid
  • Molecular formula:
    KCl in H2O

 

Frequently Bought Together