Order Entry
Austria
ContactUsLinkComponent
 
Phosphate, Ion chromatography standard solution, Specpure®, PO₄³⁻ 1000 µg/ml
 
Phosphate, Ion chromatography standard solution, Specpure®, PO₄³⁻ 1000 µg/ml
 

 

  • MDL No.:
    MFCD00145108
  • Physical form:
    Liquid
  • Odour/scent:
    Odourless
  • Molecular formula:
    KH2PO4 in H2O

 

Frequently Bought Together