Order Entry
Austria
ContactUsLinkComponent
Sulphate, Ion chromatography standard solution, Specpure®, SO₄⁻² 1000 µg/ml
Sulphate, Ion chromatography standard solution, Specpure®, SO₄⁻² 1000 µg/ml
Certificates
Sulphate, Ion chromatography standard solution, Specpure®, SO₄⁻² 1000 µg/ml
Certificates

Specifications

  • MDL No.:
    MFCD00145343
  • Physical form:
    Liquid
  • Molecular formula:
    K2SO4 in H2O

Specifications

Related Information

Frequently Bought Together